Learn More
Biotang Inc BLOTTO IN PBS 250 ML, IBB-141. 5% nonfat dry milk in PBS (10 mM Sodium phosphate, 2 mM potassium phosphate, 10 mM KCl, 137 mM NaCl), pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Catalog No. 50155958 Shop All BIOTANG Inc Products
Encompass_Preferred
BLOTTO IN PBS 250 ML, IBB-141. 5% nonfat dry milk in PBS (10 mM Sodium phosphate, 2 mM potassium phosphate, 10 mM KCl, 137 mM NaCl), pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Catalog No. 50-155-958 Supplier Biotang Inc Supplier No. BTBB734
BLOTTO IN PBS 250 ML, IBB-141. 5% nonfat dry milk in PBS (10 mM Sodium phosphate, 2 mM potassium phosphate, 10 mM KCl, 137 mM NaCl), pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.