Learn More
Bioworld Blotto (5%) in PBS, 1 L
Catalog No. 50255453
Encompass_Preferred
Blotto in PBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in PBS
Catalog No. 50-255-453 Supplier Bioworld Supplier No. 402200242
Blotto in PBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in PBS