Learn More
Bioworld Blotto in TBS, 1 L
Catalog No. 50255455
Encompass_Preferred
Blotto in TBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in TBS
Catalog No. 50-255-455 Supplier Bioworld Supplier No. 402200282
Blotto in TBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in TBS