Learn More
Biotang Inc BLOTTO in TBS 5% nonfat dry milk 0.02% Sodium azide in TBS, 250 ml
Catalog No. 50155961 Shop All BIOTANG Inc Products
Encompass_Preferred
BLOTTO in TBS 5% nonfat dry milk 0.02% Sodium azide in TBS, 250 ml.
Catalog No. 50-155-961 Supplier Biotang Inc Supplier No. BTBB737
BLOTTO in TBS 5% nonfat dry milk 0.02% Sodium azide in TBS, 250 ml.