Learn More
Santa Cruz Biotechnology BLOTTO NON-FAT DRY MILK
Supplier:  Santa Cruz Biotechnology SC2324
Encompass
Blotto, Non-fat dry milk; Lyophilized powder; Contains Santa Cruz Biotechnology immunoanalytical grade blotto; Using w/phospho-specific antibodies: add 50mM NaF to Blotto to inhibit phosphatase activity
Catalog No. NC9544655