Learn More
Santa Cruz Biotechnology BLOTTO NON-FAT DRY MILK
Catalog No. NC9730946
Encompass
Blotto non-fat dry milk Blocking Reagents for Western Blotting Applications 500 g makes a 1x solution when reconstituted to the final volume in H2O
Catalog No. NC9730946 Supplier Santa Cruz Biotechnology Supplier No. SC2325
Blotto non-fat dry milk Blocking Reagents for Western Blotting Applications 500 g makes a 1x solution when reconstituted to the final volume in H2O