Learn More
Alkali Scientific Blotto Tween 20 Blocking Solution in TBS with Azide, 1 L
SDP
Catalog No. 5000660140
Encompass_Preferred
Blotto Tween 20 Blocking Solution in TBS with Azide is a ready-to-use blocking solution used in immunoassays to prevent non-specific binding. The inclusion of Azide helps prevent microbial growth, maintaining the solution's integrity during storage and use. This formulation is particularly useful in assays like ELISA and Western Blot, where reducing background noise is crucial for obtaining accurate results. The 1-liter volume is ideal for high-throughput laboratories that require consistent blocking solutions for multiple experiments. By blocking unoccupied binding sites, this solution ensures that the target proteins bind specifically, improving the sensitivity of immunological assays.
Catalog No. 50-006-60140 Supplier Alkali Scientific Supplier No. LOT001CP
Blotto Tween 20 Blocking Solution in TBS with Azide is a ready-to-use blocking solution used in immunoassays to prevent non-specific binding. The inclusion of Azide helps prevent microbial growth, maintaining the solution's integrity during storage and use. This formulation is particularly useful in assays like ELISA and Western Blot, where reducing background noise is crucial for obtaining accurate results. The 1-liter volume is ideal for high-throughput laboratories that require consistent blocking solutions for multiple experiments. By blocking unoccupied binding sites, this solution ensures that the target proteins bind specifically, improving the sensitivity of immunological assays.