Learn More
Biotang Inc BLOTTO/TWEEN 20 IN TBS 250 ML, IBB-161. 5% nonfat dry milk, 0.05% Tween-20 in TBS (50 mM Tris-HCl and 150 mM NaCl, pH 7.4 +/- 0.2). Prepared in 18.2 megohms water and filtered through 1 micron filter.
Catalog No. 50155966 Shop All BIOTANG Inc Products
Encompass_Preferred
BLOTTO/TWEEN 20 IN TBS 250 ML, IBB-161. 5% nonfat dry milk, 0.05% Tween-20 in TBS (50 mM Tris-HCl and 150 mM NaCl, pH 7.4 +/- 0.2). Prepared in 18.2 megohms water and filtered through 1 micron filter.
Catalog No. 50-155-966 Supplier Biotang Inc Supplier No. BTBB742
BLOTTO/TWEEN 20 IN TBS 250 ML, IBB-161. 5% nonfat dry milk, 0.05% Tween-20 in TBS (50 mM Tris-HCl and 150 mM NaCl, pH 7.4 +/- 0.2). Prepared in 18.2 megohms water and filtered through 1 micron filter.