Learn More
Biotang Inc BLOTTO TWEEN IN PBS 250 ML, IBB-151, 5% nonfat dry milk with 0.05% Tween-20 in PBS (10 mM Sodium phosphate, 2 mM potassium phosphate,10 mM KCl,137 mM NaCl), 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Catalog No. 50155960 Shop All BIOTANG Inc Products
Encompass_Preferred
BLOTTO TWEEN IN PBS 250 ML, IBB-151, 5% nonfat dry milk with 0.05% Tween-20 in PBS (10 mM Sodium phosphate, 2 mM potassium phosphate,10 mM KCl,137 mM NaCl), 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Catalog No. 50-155-960 Supplier Biotang Inc Supplier No. BTBB736
BLOTTO TWEEN IN PBS 250 ML, IBB-151, 5% nonfat dry milk with 0.05% Tween-20 in PBS (10 mM Sodium phosphate, 2 mM potassium phosphate,10 mM KCl,137 mM NaCl), 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter.