Learn More
Bioworld Blotto/Tween in PBS, 500 mL
Catalog No. 50255461
Encompass_Preferred
Blotto/Antifoam in PBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.05% Tween, 0.02% Sodium azide in PBS
Catalog No. 50-255-461 Supplier Bioworld Supplier No. 402200402
Blotto/Antifoam in PBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.05% Tween, 0.02% Sodium azide in PBS