Learn More
Bioworld Blotto/Tween in TBS, 250 mL
Catalog No. 50255462
Encompass_Preferred
Blotto/Tween in TBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.05% Tween-20, 0.02% Sodium azide in TBS
Catalog No. 50-255-462 Supplier Bioworld Supplier No. 402200441
Blotto/Tween in TBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.05% Tween-20, 0.02% Sodium azide in TBS