Learn More
Research Products International Corp Blotto-Tween 20 Blocking Solution in TBS with Azide, 500 Milliliters
SDP
Catalog No. 50488567
Encompass_Preferred
Solution contains:
50mM TRIS-HCl
150mM NaCl
3% BSA
0.05% Tween-20
0.02% Sodium Azide
Optimized for use in western blotting to minimize non-specific binding of antibodies.
pH: 7.4 ± 0.2
Catalog No. 50-488-567 Supplier Research Products International Corp Supplier No. B81000500.0
Solution contains:
50mM TRIS-HCl
150mM NaCl
3% BSA
0.05% Tween-20
0.02% Sodium Azide
Optimized for use in western blotting to minimize non-specific binding of antibodies.
pH: 7.4 ± 0.2