Learn More
Grainger Fasco Centrifugal Blower, Shaded Pole, 500 Cfm, 115 V, 1 ea
Catalog No. 502763150
Encompass_Preferred
This 115 V centrifugal blower is designed for OEM replacement applications, offering reliable air movement in various HVAC systems. This unit features a shaded pole motor with thermal protection and is suitable for operation in ambient temperatures up to 40°C.
  • Output of 500 CFM
  • 2-speed operation (1,545/1,165 RPM)
  • Open air over motor enclosure
  • Class A insulation with sleeve bearings
  • Rectangular outlet with flange
  • CSA and UL listed
Catalog No. 50-276-3150 Supplier Grainger Supplier No. 161L24
This 115 V centrifugal blower is designed for OEM replacement applications, offering reliable air movement in various HVAC systems. This unit features a shaded pole motor with thermal protection and is suitable for operation in ambient temperatures up to 40°C.
  • Output of 500 CFM
  • 2-speed operation (1,545/1,165 RPM)
  • Open air over motor enclosure
  • Class A insulation with sleeve bearings
  • Rectangular outlet with flange
  • CSA and UL listed