Learn More
Cayman Chemical BLP-3 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502915540
Encompass_Preferred
An antibacterial peptide; active against N. meningitidis, N. gonorrhoeae, N. lactamica, and N. cinerea at 20 µg/ml; folds into an alpha-helix, which inserts into and disrupts bacteriomimetic membranes
Catalog No. 50-291-5540 Supplier Cayman Chemical Supplier No. 3695710 MG
An antibacterial peptide; active against N. meningitidis, N. gonorrhoeae, N. lactamica, and N. cinerea at 20 µg/ml; folds into an alpha-helix, which inserts into and disrupts bacteriomimetic membranes