Learn More
New England Biolabs, Inc. BlpI – 500 units
SDP
Catalog No. 50812292
Encompass
A restriction endonuclease that recognizes the sequence GC^TNA_GC.
Catalog No. 50-812-292 Supplier New England Biolabs, Inc. Supplier No. R0585S
A restriction endonuclease that recognizes the sequence GC^TNA_GC.