Learn More
Grainger BLS1255 Overlaminate 50 ft L
Catalog No. 502752215
Encompass_Preferred
The Brady Laminating Roll Cold Lamination, 14A462 is a 1.5 mil thick laminating film that provides document protection from moisture, handling and chemicals. It has a gloss finish and comes in a roll that is 2 inches wide and 25 feet long. This laminating film is ideal for protecting labels, tags, signs, menus and identification badges. It applies at room temperature using a cold process and works with thermal transfer, inkjet and laser printed documents. The laminate is UL recognized and offers excellent clarity for highlighting printed information. Overall, this is a high quality laminating film for protecting important documents from wear.
Catalog No. 50-275-2215 Supplier Grainger Supplier No. 14A462
The Brady Laminating Roll Cold Lamination, 14A462 is a 1.5 mil thick laminating film that provides document protection from moisture, handling and chemicals. It has a gloss finish and comes in a roll that is 2 inches wide and 25 feet long. This laminating film is ideal for protecting labels, tags, signs, menus and identification badges. It applies at room temperature using a cold process and works with thermal transfer, inkjet and laser printed documents. The laminate is UL recognized and offers excellent clarity for highlighting printed information. Overall, this is a high quality laminating film for protecting important documents from wear.