Learn More
Cayman Chemical BLT1 RECPTR Blockng PeptId 1ea
SDP
Catalog No. 502955824
Encompass_Preferred
A purine derivative that reversibly antagonizes aryl hydrocarbon receptor signaling (IC50 127 nM in CD34 cells) induces a 50-fold increase in human embryonic stem cells expressing CD34 (EC50 120 nM) and a 17-fold increase in stem cells that retain the ability to engraft immunodeficient mice
Catalog No. 50-295-5824 Supplier Cayman Chemical Supplier No. 1201121 EA
A purine derivative that reversibly antagonizes aryl hydrocarbon receptor signaling (IC50 127 nM in CD34 cells) induces a 50-fold increase in human embryonic stem cells expressing CD34 (EC50 120 nM) and a 17-fold increase in stem cells that retain the ability to engraft immunodeficient mice