Learn More
Cayman Chemical BLT2 Receptor Polyclonal Antibody, 1 ea
SDP
Catalog No. 502952908
Encompass_Preferred
Immunogen: peptide from the C-terminal region of human BLT2 • Host: Rabbit • Species Reactivity: (+) Human and mouse • Application(s): WB
Catalog No. 50-295-2908 Supplier Cayman Chemical Supplier No. 1201241 EA
Immunogen: peptide from the C-terminal region of human BLT2 • Host: Rabbit • Species Reactivity: (+) Human and mouse • Application(s): WB