Learn More
Cayman Chemical BLT2 RECPTR Blockng PeptId 1ea
SDP
Catalog No. 502949140
Encompass_Preferred
A ganglioside GD1b molecular species isolated from porcine brain with primarily C180 fatty acyl chain lengths a component of plasma membranes packs densely with cholesterol to form lipid microdomains that modulate both intra- and intercellular signaling events the concentration of ganglioside GD1b in human brain increases with age
Catalog No. 50-294-9140 Supplier Cayman Chemical Supplier No. 3201241 EA
A ganglioside GD1b molecular species isolated from porcine brain with primarily C180 fatty acyl chain lengths a component of plasma membranes packs densely with cholesterol to form lipid microdomains that modulate both intra- and intercellular signaling events the concentration of ganglioside GD1b in human brain increases with age