Learn More
Selleck Chemical LLC BLU 451-E1877-1G
SDP
Catalog No. 503135882
Encompass_Preferred
BLU 451 is a central nervous system (CNS)-penetrant wild-type EGFR-sparing covalent inhibitor of epidermal growth factor receptor (EGFR) exon 20 insertion (Ex20ins) with an IC50 of 7-78 nM It also exhibits a potent anti-tumor efficacy
Catalog No. 50-313-5882 Supplier Selleck Chemical LLC Supplier No. E18771G
BLU 451 is a central nervous system (CNS)-penetrant wild-type EGFR-sparing covalent inhibitor of epidermal growth factor receptor (EGFR) exon 20 insertion (Ex20ins) with an IC50 of 7-78 nM It also exhibits a potent anti-tumor efficacy