Learn More
Apexbio Technology LLC BLU-554, 50mg. Cas: 1707289-21-1 MFCD: MFCD30496704.
SDP
Supplier:  Apexbio Technology LLC B839150
Encompass_Preferred
BLU-554 is an effective, highly selective, orally active FGFR4 inhibitor with IC50 of 5 nM. The IC50 range for FGFR1-3 is 624-2203 nM. In vitro, BLU-554 interacts with FGFR4 and has high potency and selectivity for FGFR4 inhibition; in vivo, BLU-554 is well tolerated in tumor models and induces tumor regression in liver cancer models. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8430