Learn More
Selleck Chemical LLC Fisogatinib (BLU-554) 2mg 1707289-21-1
SDP
Supplier:  Selleck Chemical LLC S85032MG
Encompass_Preferred
Fisogatinib (BLU-554) is a potent, highly-selective, oral FGFR4 inhibitor with an IC50 value of 5 nM. The IC50s for FGFR1-3 is 624-2203 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6095