Learn More
Medchemexpress LLC BLU-945 | 2660250-10-0 | 99.4% | C28H37FN6O3S | 100 MG
SDP
Catalog No. 5000355348
Encompass_Preferred
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation, making it suitable for research into lung cancer, including non-small cell lung cancer (NSCLC).
  • Potent, highly selective, reversible, and orally active EGFR tyrosine kinase inhibitor.
  • Effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation.
  • Used for research of lung cancer, including non-small cell lung cancer (NSCLC).
  • Exhibits inhibitory activity against EGFRm/T790M double and EGFRm/T790M/C797S triple mutants.
  • Demonstrates potent, robust EGFR pathway inhibition and anti-tumor activity in Osimertinib-resistant models.
Catalog No. 50-003-55348 Supplier Medchemexpress LLC Supplier No. HY144680100MG
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation, making it suitable for research into lung cancer, including non-small cell lung cancer (NSCLC).
  • Potent, highly selective, reversible, and orally active EGFR tyrosine kinase inhibitor.
  • Effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation.
  • Used for research of lung cancer, including non-small cell lung cancer (NSCLC).
  • Exhibits inhibitory activity against EGFRm/T790M double and EGFRm/T790M/C797S triple mutants.
  • Demonstrates potent, robust EGFR pathway inhibition and anti-tumor activity in Osimertinib-resistant models.