Learn More
Medchemexpress LLC BLU-945 | 2660250-10-0 | 99.4% | C28H37FN6O3S | 25 MG
SDP
Catalog No. 5000357144
Encompass_Preferred
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKIs). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation. BLU-945 can be used for research related to lung cancer, including non-small cell lung cancer (NSCLC).
Catalog No. 50-003-57144 Supplier Medchemexpress LLC Supplier No. 25MGHY144680
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKIs). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation. BLU-945 can be used for research related to lung cancer, including non-small cell lung cancer (NSCLC).