Learn More
Medchemexpress LLC Blu-945 | 2660250-10-0 | 99.6% | C28H37FN6O3S | 1 ML
SDP
Catalog No. 5000354817
Encompass_Preferred
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKIs). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation, making it suitable for lung cancer research, including non-small cell lung cancer (NSCLC).
  • Potent, highly selective, reversible, and orally active
  • Inhibits EGFR with L858R, exon 19 deletion, T790M, and C797S mutations
  • Used for research of lung cancer, including non-small cell lung cancer (NSCLC)
  • Demonstrates anti-tumor activity in triple-mutant Osimertinib-resistant models
Catalog No. 50-003-54817 Supplier Medchemexpress LLC Supplier No. HY14468010MM/1ML
BLU-945 is a potent, highly selective, reversible, and orally active epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKIs). It effectively inhibits EGFR with L858R and/or exon 19 deletion mutation, T790M mutation, and C797S mutation, making it suitable for lung cancer research, including non-small cell lung cancer (NSCLC).
  • Potent, highly selective, reversible, and orally active
  • Inhibits EGFR with L858R, exon 19 deletion, T790M, and C797S mutations
  • Used for research of lung cancer, including non-small cell lung cancer (NSCLC)
  • Demonstrates anti-tumor activity in triple-mutant Osimertinib-resistant models