Learn More
TARGETMOL CHEMICALS INC BLU-945 5MG
SDP
Catalog No. 502578965
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BLU-945 is a potent highly selective reversible and orally active inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase. BLU-945 can effectively inhibit EGFR with L858R and/or exon 19 deletion mutation T790M mutation and C797S mutation. BLU-945 can be used for the research of lung cancer including non-small cell lung cancer (NSCLC). purity: 99%
Catalog No. 50-257-8965 Supplier TARGETMOL CHEMICALS INC Supplier No. T97545MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BLU-945 is a potent highly selective reversible and orally active inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase. BLU-945 can effectively inhibit EGFR with L858R and/or exon 19 deletion mutation T790M mutation and C797S mutation. BLU-945 can be used for the research of lung cancer including non-small cell lung cancer (NSCLC). purity: 99%