Learn More
Selleck Chemical LLC BLU-945-E1176-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC E117610MM/1ML
Encompass_Preferred
BLU-945 is a potent highly selective reversible and orally active epidermal growth factor receptor(EGFR) tyrosine kinase inhibitor (TKIs) BLU-945 can be used for the research of lung cancer including non-small cell lung cancer (NSCLC)
Catalog No. 50-313-7973