Learn More
Selleck Chemical LLC BLU-945-E1176-5MG
SDP
Catalog No. 503138224
Encompass_Preferred
BLU-945 is a potent highly selective reversible and orally active epidermal growth factor receptor(EGFR) tyrosine kinase inhibitor (TKIs) BLU-945 can be used for the research of lung cancer including non-small cell lung cancer (NSCLC)
Catalog No. 50-313-8224 Supplier Selleck Chemical LLC Supplier No. E11765MG
Add to Cart
Add to Cart
BLU-945 is a potent highly selective reversible and orally active epidermal growth factor receptor(EGFR) tyrosine kinase inhibitor (TKIs) BLU-945 can be used for the research of lung cancer including non-small cell lung cancer (NSCLC)