Learn More
Ted Pella Inc Cryogenic Tape, 19mm x 15M, Roll, Blue
SDP
Catalog No. NC1874216
Encompass
NitroTAPE is a permanent cryogenic tape for plastic and glass vials, microtubes, cryo and freezer boxes, plastic bags and other containers. Intended for long-term specimen preservation in liquid phase and vapor phase liquid nitrogen (-196°C), laboratory freezers (-20°C, -70°C, -80°C, -120°C) and dry ice (-78.5°C).
Catalog No. NC1874216 Supplier Ted Pella Inc Supplier No. 223162
NitroTAPE is a permanent cryogenic tape for plastic and glass vials, microtubes, cryo and freezer boxes, plastic bags and other containers. Intended for long-term specimen preservation in liquid phase and vapor phase liquid nitrogen (-196°C), laboratory freezers (-20°C, -70°C, -80°C, -120°C) and dry ice (-78.5°C).