Learn More
Bellco Glass, Inc. Blu S/Cap W/PTFE/Sil Septa,9mm 9-425
SDP
Catalog No. 501945385
Encompass_Preferred
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 9mm
Catalog No. 50-194-5385 Supplier Bellco Glass, Inc. Supplier No. 207000950
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 9mm