Learn More
TARGETMOL CHEMICALS INC BLU0588 5MG
SDP
Catalog No. 502579655
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BLU0588 is a selective PRKACA inhibitor. BLU0588 inhibits PRKACA catalytic activity with a half-maximal inhibitory concentration (IC50) of 1 nM.
Catalog No. 50-257-9655 Supplier TARGETMOL CHEMICALS INC Supplier No. T60169
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BLU0588 is a selective PRKACA inhibitor. BLU0588 inhibits PRKACA catalytic activity with a half-maximal inhibitory concentration (IC50) of 1 nM.