Learn More
Medchemexpress LLC Blu2864 | 2810747-89-6 | 99.5% | 452.43 | 100 MG
SDP
Catalog No. 5000361062
Encompass_Preferred
BLU2864 is an orally active, highly selective, ATP-competitive PRKACA inhibitor with an IC50 of 0.3 nM for PRKACA. It exhibits anti-tumor activity and can be utilized in cancer and polycystic kidney disease research.
  • Inhibits forskolin-induced in vitro cystogenesis
  • Suppresses renal basal and total PKA activities
  • Leads to higher urine outputs and lower kidney weights
  • Reduces FLC tumor growth
Catalog No. 50-003-61062 Supplier Medchemexpress LLC Supplier No. HY150076100MG
BLU2864 is an orally active, highly selective, ATP-competitive PRKACA inhibitor with an IC50 of 0.3 nM for PRKACA. It exhibits anti-tumor activity and can be utilized in cancer and polycystic kidney disease research.
  • Inhibits forskolin-induced in vitro cystogenesis
  • Suppresses renal basal and total PKA activities
  • Leads to higher urine outputs and lower kidney weights
  • Reduces FLC tumor growth