Learn More
TARGETMOL CHEMICALS INC BLU9931 25MG
SDP
Catalog No. 502577151
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BLU9931 is the first selective small molecule inhibitor of FGFR4. purity: 99%
Catalog No. 50-257-7151 Supplier TARGETMOL CHEMICALS INC Supplier No. T234725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BLU9931 is the first selective small molecule inhibitor of FGFR4. purity: 99%