Learn More
Selleck Chemical LLC BLU9931-5mg
SDP
Catalog No. 502390063
Encompass_Preferred
BLU9931 is a potent, selective, and irreversible FGFR4 inhibitor with IC50 of 3 nM, about 297-, 184-, and 50-fold selectivity over FGFR1/2/3, respectively.
Catalog No. 50-239-0063 Supplier Selleck Chemical LLC Supplier No. S78195MG
BLU9931 is a potent, selective, and irreversible FGFR4 inhibitor with IC50 of 3 nM, about 297-, 184-, and 50-fold selectivity over FGFR1/2/3, respectively.