Learn More
Andwin Scientific Laser Cryo-Tags 1.69 x 0.75", Blue, 1,040/pk
SDP
Catalog No. NC1662610
Encompass_Preferred
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.
Catalog No. NC1662610 Supplier Andwin Scientific Supplier No. LCRY1100B
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.