Learn More
Analytical Sales & Services Blue 100µL Skirted Low Profile Plate
Catalog No. 5000804375
Encompass
Advantage Blue 100µL Skirted Low Profile Plate. Impermeable to Water Vapor.\n\nQty/Pkg: 100
Catalog No. 50-008-04375 Supplier Analytical Sales & Services Supplier No. 96100B
Advantage Blue 100µL Skirted Low Profile Plate. Impermeable to Water Vapor.\n\nQty/Pkg: 100