Learn More
Analytical Sales & Services Blue 200µL PCR Plate - Flat Well Rim, No Skirt
Supplier:  Analytical Sales & Services 969610B
Encompass
Blue 200µL PCR Plate Flat Well Rim Thin Walled - Can Be Cut Into Sections\n\nQty/Pkg: 100
Catalog No. 50-008-04416