Learn More
Alkali Scientific 5mL Centrifuge Tubes, 4 Bags of 50, Blue, 200/cs
SDP
Catalog No. 502388825
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 200 blue 5mL centrifuge tubes.
Catalog No. 50-238-8825 Supplier Alkali Scientific Supplier No. C3005B
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 200 blue 5mL centrifuge tubes.