Learn More
RIVERDALE SCIENTIFIC LLC BLUE -ASPIRATOR ADAPTER FOR 1 ML PIPETTE TIP, ALTERNATIVE TO Glass PASTEUR PIPETTE, USE WITH VACBUDDY, 5/pack
Catalog No. NC2884360
Encompass_Preferred
These ASPIRATOR ADAPTER tips can conveniently be used to aspirate out old cell culture media; add disposable sterile 1ml pipette tip. Available in pack of 5. Autoclavable. Can also be used for:•Aspirating aqueous or organic phases.•Aspirating body fluids or wash fluids during animal dissections or surgeries.•The adapter serves as alternative to glass Pasteur pipettes. It fits standard 1 ml pipette tips that are readily available in research labs. •It is suitable for use with 1/4th inch ID tubing.
Catalog No. NC2884360 Supplier RIVERDALE SCIENTIFIC LLC Supplier No. RSBLUDPT
These ASPIRATOR ADAPTER tips can conveniently be used to aspirate out old cell culture media; add disposable sterile 1ml pipette tip. Available in pack of 5. Autoclavable. Can also be used for:•Aspirating aqueous or organic phases.•Aspirating body fluids or wash fluids during animal dissections or surgeries.•The adapter serves as alternative to glass Pasteur pipettes. It fits standard 1 ml pipette tips that are readily available in research labs. •It is suitable for use with 1/4th inch ID tubing.