Learn More
Mopec Blue Biopsy Sponges
SDP
Catalog No. 22444387 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
1.25 x 1 in.
Catalog No. Dimensions (L x W)
22-444-387 1.25 x 1 in.
1 options
Catalog No. 22-444-387 Supplier Mopec Supplier No. BI056
Add to Cart
Add to Cart
Biopsy Pads stand out for their reliable specimen support, high visibility, and compatibility with standard cassettes, making them a dependable, cost-effective choice for routine use in pathology and histology laboratories.
  • Quantity: 1000 papers per box
  • Specimen stabilization: Holds small biopsy tissue securely in place within cassettes during handling, transfer, and processing, reducing the risk of specimen movement, loss, or damage.
  • Cushioning protection: The soft foam material helps prevent compression and pressure-related artifacts, preserving tissue integrity during processing.
  • High visibility: The blue color provides excellent contrast against most tissue types, aiding quick visual identification during embedding and sectioning.
  • Efficient reagent flow: The open-cell foam structure allows fixatives and processing reagents to flow freely around the specimen, supporting proper absorption.
Specifications
Product Type Biopsy Pad
Material Polyester Urethane Foam
Color Blue
Dimensions (L x W) 1.25 x 1 in.
Disposable Single-use