Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Blue Biopsy Wraps
SDP

Catalog No. 22444388 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
1.5 x 2 in.
2.5 x 4 in.
2 x 2.5 in.
2 x 3 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-444-388 1.5 x 2 in.
22-444-389 2 x 2.5 in.
22-444-390 2 x 3 in.
22-444-391 2.5 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-444-388 Supplier Mopec Supplier No. BI057
Add to Cart
Add to Cart

Blue Biopsy Wraps are high-quality, disposable biopsy specimen wraps designed to securely hold and protect small tissue samples during laboratory processing. These wraps are available in multiple sizes to accommodate a range of specimen sizes and preferences in workflow.

  • Quantity: 500 papers per box
  • Specimen protection: Helps contain and maintain the identity of small biopsy tissue during handling, transfer, and processing, reducing the risk of loss or damage.
  • High visibility: The blue color provides excellent contrast against most tissue types and paraffin, aiding quick visual identification during embedding and sectioning.
  • Chemical resistance: The lens paper construction resists common histology reagents such as water, alcohol, and xylene, ensuring the wrap doesn't deteriorate or interfere as specimens move through processing steps.
  • Flexible sizing: Multiple sizes allow labs to choose the best fit for different biopsy dimensions, improving efficiency and reducing waste.

Specifications

Product Type Biopsy Paper
Material Lens Paper
Color Blue
Dimensions (L x W) 1.5 x 2 in.
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.