Learn More
Leica Biosystems BLUE CHROMOGEN
Catalog No. NC1940150
Encompass
Blue Chromogen is designed to be used in conjunction with DS9914 BOND Polymer Refine HRP Plex Detection kit. This chromogen provides a vibrant blue color and is suitable for use with aqueous mounting media. Our Blue Chromogen provides brilliant contrast against other chromogens and counterstains including hematoxylin. Take advantage of on-board chromogen mixing on BOND instruments, not to mention the fully automated staining opportunities using BOND detection systems and BOND ready-to-use primary antibodies and probes.
Catalog No. NC1940150 Supplier Leica Biosystems Supplier No. DC9896
Blue Chromogen is designed to be used in conjunction with DS9914 BOND Polymer Refine HRP Plex Detection kit. This chromogen provides a vibrant blue color and is suitable for use with aqueous mounting media. Our Blue Chromogen provides brilliant contrast against other chromogens and counterstains including hematoxylin. Take advantage of on-board chromogen mixing on BOND instruments, not to mention the fully automated staining opportunities using BOND detection systems and BOND ready-to-use primary antibodies and probes.