Learn More
MP Biomedicals™ Blue Clickmaster Ratchet Nut for FastPrep-24™ 5G and Classic
Shop All MP Biomedicals, Inc ProductsDescription
- The FastPrep™ is the only available homogenizer with multiple interchangeable adapters which allows a unique versatility in sample number, size, and temperature conditions. All optional adapters stand stable on the benchtop and are commonly used as tube racks for sample storage at -20°C or -80°C.
- The securing knob is important for maintaining the tube holder adapters on the central axis or during the 3-step assembly of the FastPrep-24™.
- One Blue Clickmaster Ratchet Nut supports up to 11 different adapters. From lightweight ones to heavy and sturdy adapters, the Blue Clickmaster provides the easiest and most secure locking mechanism. Thanks to its high torque, it is particularly recommended for the heaviest adapters and especially for metal tube holders.
- This product can be used on FastPrep-24™Classic or FastPrep-24™ 5G
Specifications
Specifications
| Blade Material | Stainless Steel |
| Closure Color | Blue |
| For Use With (Equipment) | FastPrep-245G and Classic |
| Product Type | Ratchet Nut |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.