Learn More
MSC 80/20 Inc. Button Head Socket Cap Screw, Steel, M8 x 1.25, 20.00 mm, 1 ea
Catalog No. 5000512194
Encompass
This button head socket cap screw is a reliable steel fastener designed for secure assembly in various industrial applications.
  • Features a durable steel construction with a zinc finish
  • Designed for use with 15, 30, and 40 series systems
  • Includes full thread M8 x 1.25 threading
  • Measures 20.00 mm in total length
  • Rated as ESD safe
Catalog No. 50-005-12194 Supplier MSC Supplier No. 54976683
This button head socket cap screw is a reliable steel fastener designed for secure assembly in various industrial applications.
  • Features a durable steel construction with a zinc finish
  • Designed for use with 15, 30, and 40 series systems
  • Includes full thread M8 x 1.25 threading
  • Measures 20.00 mm in total length
  • Rated as ESD safe