Learn More
Sigma Aldrich Fine Chemicals Biosciences Blue Dextran, 87915-38-6, MFCD00130661, 5 g
Catalog No. NC0056096
Encompass
Blue dextran is used in affinity chromatography, gel filtration chromatography, protein chromatography and molecular weight markers. Blue dextran has been used to study Alzheimer′s disease. Void volume marker for gel filtration columns, Synonym: Dextran blue from Leuconostoc mesenteroides.
Catalog No. NC0056096 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D57515G
Blue dextran is used in affinity chromatography, gel filtration chromatography, protein chromatography and molecular weight markers. Blue dextran has been used to study Alzheimer′s disease. Void volume marker for gel filtration columns, Synonym: Dextran blue from Leuconostoc mesenteroides.