Learn More
Medchemexpress LLC Blue dextran | 87915-38-6 | MFCD00130661 | 2,000,000 Da | (C6H10O5)n | 250 MG
SDP
Catalog No. 5000283567
Encompass_Preferred
Blue dextran (MW 2,000,000) is a high-molecular-weight dextran conjugated to a blue dye, supplied as a solid for research use. It functions as a large, water-soluble macromolecular probe used in biochemical assays and chromatographic separations where visualization or exclusion of high-mass species is required.
  • High molecular weight (≈2,000,000 Da) tracer for size-exclusion and gel-filtration studies.
  • Water-soluble, colored conjugate that enables visual detection.
  • Useful as a column void volume and macromolecule exclusion marker.
  • Supplied as a solid for easy storage and preparation.
  • Suitable for research applications requiring non-reactive, high-mass probes.
Catalog No. 50-002-83567 Supplier Medchemexpress LLC Supplier No. HYW134328250MG
Blue dextran (MW 2,000,000) is a high-molecular-weight dextran conjugated to a blue dye, supplied as a solid for research use. It functions as a large, water-soluble macromolecular probe used in biochemical assays and chromatographic separations where visualization or exclusion of high-mass species is required.
  • High molecular weight (≈2,000,000 Da) tracer for size-exclusion and gel-filtration studies.
  • Water-soluble, colored conjugate that enables visual detection.
  • Useful as a column void volume and macromolecule exclusion marker.
  • Supplied as a solid for easy storage and preparation.
  • Suitable for research applications requiring non-reactive, high-mass probes.