Learn More
Medchemexpress LLC Blue dextran | 87915-38-6 | MFCD00130661 | 5,000 Da | [C6H10O5]n | 250 MG
SDP
Catalog No. 5000283460
Encompass_Preferred
Blue dextran (MW 5000) is a dextran polymer covalently conjugated to the Cibacron Blue 3G-A ligand, supplied as a powdered reagent. It functions as an affinity ligand and tracer in chromatographic and biochemical workflows, commonly used for purification and analysis of serum albumin and as a molecular-weight marker in gel filtration applications.
  • Cibacron Blue 3G-A conjugated to dextran for reversible protein binding.
  • Nominal molecular weight 5,000 Da suitable for gel filtration tracers.
  • Provided as a dry powder for easy dissolution in aqueous buffers.
  • Suitable for affinity purification of human serum albumin and related proteins.
  • Available in milligram pack sizes for laboratory use.
Catalog No. 50-002-83460 Supplier Medchemexpress LLC Supplier No. HYW134328A250MG
Blue dextran (MW 5000) is a dextran polymer covalently conjugated to the Cibacron Blue 3G-A ligand, supplied as a powdered reagent. It functions as an affinity ligand and tracer in chromatographic and biochemical workflows, commonly used for purification and analysis of serum albumin and as a molecular-weight marker in gel filtration applications.
  • Cibacron Blue 3G-A conjugated to dextran for reversible protein binding.
  • Nominal molecular weight 5,000 Da suitable for gel filtration tracers.
  • Provided as a dry powder for easy dissolution in aqueous buffers.
  • Suitable for affinity purification of human serum albumin and related proteins.
  • Available in milligram pack sizes for laboratory use.