Learn More
Medchemexpress LLC Blue dextran | 87915-38-6 | MFCD00130661 | 70,000 Da | 250 MG
SDP
Catalog No. 5000283477
Encompass_Preferred
Blue dextran (average mol wt 70,000 Da) is a dextran polymer conjugated to the Cibacron Blue 3G-A ligand, supplied as a light blue solid and intended for biochemical applications such as affinity purification and chromatographic void-volume marking. Batch-specific certificates of analysis report physical characteristics and substitution metrics to support experimental use.
  • Conjugated dextran-Cibacron Blue ligand suitable for affinity purification.
  • Useful as a void-volume marker and for albumin purification.
  • Light blue solid form that is easy to handle and weigh.
  • Supplied with certificate of analysis reporting loss on drying and degree of substitution.
  • Average molecular weight ~70,000 Da for size-dependent applications.
Catalog No. 50-002-83477 Supplier Medchemexpress LLC Supplier No. HYW134328E250MG
Add to Cart
Add to Cart
Blue dextran (average mol wt 70,000 Da) is a dextran polymer conjugated to the Cibacron Blue 3G-A ligand, supplied as a light blue solid and intended for biochemical applications such as affinity purification and chromatographic void-volume marking. Batch-specific certificates of analysis report physical characteristics and substitution metrics to support experimental use.
  • Conjugated dextran-Cibacron Blue ligand suitable for affinity purification.
  • Useful as a void-volume marker and for albumin purification.
  • Light blue solid form that is easy to handle and weigh.
  • Supplied with certificate of analysis reporting loss on drying and degree of substitution.
  • Average molecular weight ~70,000 Da for size-dependent applications.