Learn More
USA Scientific Inc Direct Thermal Combo Cryo-Tags on rolls, blue
Supplier:  USA Scientific Inc 91386001
Encompass
Direct Thermal Combo Cryo-Tags, blue, 1.5 inch L x 0.5 inch W and 3/8 inch diameter, 500 each/roll. For direct thermal printers.
Catalog No. NC0500196