Learn More
BURKLE INC Blue disposable liquid funnel PS, Ø 3.94 in. (100 mm), sterile
Catalog No. 503081353
Encompass_Preferred
The blue disposable polystyrene funnel for liquids is produced in a Class 7 clean room and sterilized individually for safe single-use.
Its unique blue color doesn’t occur naturally in food, allowing quick visual detection if contamination occurs.
This supports HACCP compliance by preventing foreign object hazards, crucial in food, animal feed, and pharmaceutical industries.
Ideal for filling bottles and containers with narrow openings, it ensures no liquid or solid loss during transfer.
The funnel features a suspension eye and venting fins for smooth, uniform flow while pouring.
Perfect for sampling, lab use, and filtering when combined with filter paper, this hygienic tool offers reliable liquid handling.
Catalog No. 50-308-1353 Supplier BURKLE INC Supplier No. 53786003
The blue disposable polystyrene funnel for liquids is produced in a Class 7 clean room and sterilized individually for safe single-use.
Its unique blue color doesn’t occur naturally in food, allowing quick visual detection if contamination occurs.
This supports HACCP compliance by preventing foreign object hazards, crucial in food, animal feed, and pharmaceutical industries.
Ideal for filling bottles and containers with narrow openings, it ensures no liquid or solid loss during transfer.
The funnel features a suspension eye and venting fins for smooth, uniform flow while pouring.
Perfect for sampling, lab use, and filtering when combined with filter paper, this hygienic tool offers reliable liquid handling.